• USA Home
  • 55501 - (5R)-5-Hydroxy-L-lysine dihydrochloride monohydrate

EMAIL THIS PAGE TO A FRIEND
55501 Aldrich

(5R)-5-Hydroxy-L-lysine dihydrochloride monohydrate

≥99.0% (AT)

Synonym: (2S,5R)-2,6-Diamino-5-hydroxycaproic acid dihydrochloride

  • CAS Number 355118-29-5

  • Empirical Formula (Hill Notation) C6H14N2O3 · 2HCl · H2O

  • Molecular Weight 253.12

  •  Beilstein Registry Number 8010462

  •  MDL number MFCD00210056

  •  PubChem Substance ID 57651501

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Lysine Derivatives, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   AQDHDEQAXDPVJQ-LEIZOONBSA-N
assay   ≥99.0% (AT)
optical activity   [α]20/D +11.5±1.0°, c = 1% in 6 M HCl

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?