• USA Home
  • 557382 - 4′-Chloro-5-fluoro-2-hydroxybenzophenone

EMAIL THIS PAGE TO A FRIEND
557382 Aldrich

4′-Chloro-5-fluoro-2-hydroxybenzophenone

96%

Purchase

Properties

Related Categories Building Blocks, C13 to C14, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   AYBQWBCUAWOLCT-UHFFFAOYSA-N
assay   96%
mp   65-69 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?