• USA Home
  • 558494 - 4-(4-Nitrophenyl)-1,2,3-thiadiazole

EMAIL THIS PAGE TO A FRIEND
558494 Aldrich

4-(4-Nitrophenyl)-1,2,3-thiadiazole

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Thiadiazoles
InChI Key   VLRKUBATYOJPSP-UHFFFAOYSA-N
assay   97%
mp   207-211 °C(lit.)

Description

Packaging

1, 5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?