• USA Home
  • 558923 - 2-(Phenylsulfonyl)-1,3-cycloheptadiene

EMAIL THIS PAGE TO A FRIEND
558923 Aldrich

2-(Phenylsulfonyl)-1,3-cycloheptadiene

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfones, Sulfur Compounds More...
InChI Key   JBFCGKYPRPLNFA-UHFFFAOYSA-N
assay   97%
mp   60-64 °C(lit.)

Description

Packaging

1 g in glass bottle

Application

2-(Phenylsulfonyl)-1,3-cycloheptadiene may be used to synthesize 7-ring vinyl sulfone epoxide, via Jacobsen catalytic asymmetric epoxidation.

General description

2-(Phenylsulfonyl)-1,3-cycloheptadiene can be synthesized from 1,3-cycloheptadiene and phenyl benzeneselenosulfonate. Achiral 2-(phenylsulfonyl)-1,3-cycloheptadiene has been obtained from cycloheptanone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?