• USA Home
  • 560588 - PSS-Octa(2-trichlorosilylethyl) substituted

EMAIL THIS PAGE TO A FRIEND
560588 Aldrich

PSS-Octa(2-trichlorosilylethyl) substituted

Synonym: 1,3,5,7,9,11,13,15-Octa(2-trichlorosilyl)ethyl)pentacyclo-[9.5.1.13,9.15,15.17,13]octasiloxane, Octa(trichlorosilylethyl)-POSS®, POSS®-Octa(trichlorosilylethyl) substituted

Purchase

Properties

Related Categories Completely Condensed, Materials Science, Nanomaterials, Silsesquioxanes: POSS Nanohybrids
InChI Key   VRQHRTGJRVVVRD-UHFFFAOYSA-N
mp   >300 °C(lit.)
solubility   THF: soluble
  acetonitrile: insoluble
  chloroform: soluble

Description

Packaging

5 g in glass bottle

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?