• USA Home
  • 560618 - 2-[1-(Hydroxymethyl)ethyl] oxazoline

EMAIL THIS PAGE TO A FRIEND
560618 Aldrich

2-[1-(Hydroxymethyl)ethyl] oxazoline

Synonym: 2-[1-(Hydroxymethyl)ethyl]-4,5-dihydrooxazole

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Oxazolines/Oxazolidines
InChI Key   SWYMJLYPXAXHBJ-UHFFFAOYSA-N
refractive index   n20/D 1.4800(lit.)
density   1.1240 g/mL at 25 °C(lit.)

Description

Application

Precursor to 2-Isopropenyl-2-oxazoline (IPO)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?