• USA Home
  • 560677 - 2,5-Bis(3′,7′-dimethyloctyloxy)benzene-1,4-diacetonitrile

EMAIL THIS PAGE TO A FRIEND
560677 Aldrich

2,5-Bis(3′,7′-dimethyloctyloxy)benzene-1,4-diacetonitrile

98%

  • CAS Number 480438-86-6

  • Linear Formula [(CH3)2CH(CH2)3CH(CH3)CH2CH2O]2C6H2(CH2CN)2

  • Molecular Weight 468.71

  •  MDL number MFCD03701546

  •  PubChem Substance ID 24879885

Purchase

Properties

Related Categories Materials Science, Organic and Printed Electronics, Polyphenylenevinylene (PPV, CN-PPV) Monomers, Synthetic Tools and Reagents
InChI Key   KGJJVMVPAVTWCT-UHFFFAOYSA-N
assay   98%
mp   83.0-87.9 °C(lit.)

Description

General description

Monomeric precursor for the light emitting polymer OC10C10-CN-PPV

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?