• USA Home
  • 560928 - 5-(5-Chloro-2-hydroxyphenyl)-1-phenylpyrazole acetate

EMAIL THIS PAGE TO A FRIEND
560928 Aldrich

5-(5-Chloro-2-hydroxyphenyl)-1-phenylpyrazole acetate

97%

Purchase

Properties

Related Categories Building Blocks, C10-C30+, C12 to C37, Chemical Synthesis, Halogenated Heterocycles,
InChI Key   IGLQLOAOZMXFPO-UHFFFAOYSA-N
assay   97%
mp   147-150 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?