• USA Home
  • 561134 - Polyaniline (emeraldine salt) long chain, grafted to lignin

EMAIL THIS PAGE TO A FRIEND
561134 Aldrich

Polyaniline (emeraldine salt) long chain, grafted to lignin

powder

Purchase

Properties

Related Categories Emeraldine Salts, Materials Science, Materials for Buffer Layer, Organic Photovoltaic (OPV) Materials, Organic and Printed Electronics,
form   powder
contains   ligno-sulfonic acid and para-toluene sulfonic acid as dopant
particle size   2-3 μm
pH   1.9 (5 wt. % in H2O)
conductivity   4-6 S/cm

Description

Chem/Phys Resistance

Stable at 300°C for 30 mins. Insoluble in most solvents. Dispersible over a wide pH range in water and polar, protic organic solvents including isopropanol, DMSO, DMF, and NMP.

Features and Benefits

Inherently conductive polymer. Redox active up to a pH of 9.

Other Notes

Shake well before use.

Application

Additive in polymer blends and liquid dispersions for corrosion resistant coatings; antistatic fabrics, coatings, and packaging; conductive inks and adhesives; printed circuit boards; radar and microwave absorption; EMI/RFI shielding; sensors.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Lithography Nanopatterning Tutorial

Lithography Nanoimprint Lithography Soft Lithography Photochemical Acid Generators Nanopatterning Nanopatterning via Phase Separation of Polymers Self-Assembled Monolayer Systems (SAMS) Norbornadiene...
Keywords: Adhesion, Adsorption, Amplification, Asymmetric synthesis, Biomaterials, Building blocks, Catalysis, Cycloadditions, Deposition, Eliminations, Evaporation, Hydrogenations, Isomerizations, Materials Science, Microelectronics, Petrochemical, Pharmaceutical, Rearrangements, Ring opening metathesis polymerisation, Semiconductor, Separation, Solvents, Spin coating, Thin film deposition, transformation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?