• USA Home
  • 561231 - Poly(4-styrenesulfonic acid) lithium salt solution

EMAIL THIS PAGE TO A FRIEND
561231 Aldrich

Poly(4-styrenesulfonic acid) lithium salt solution

Mw ~75,000, 30 wt. % in H2O

Synonym: PSS, Poly(4-lithium styrenesulfonic acid) solution

Purchase

Properties

Related Categories Anionic Polyelectrolytes - PSSA & Others, Hydrophilic Polymers, Materials Science, Micro/NanoElectronics, Poly(styrenesulfonate) (PSS),
mol wt   Mw ~75,000
concentration   30 wt. % in H2O
refractive index   n20/D 1.3985
pH   4-6 (25 °C)
viscosity   100-300 cP(25 °C, Brookfield) (20 rpm)
density   1.12 g/mL at 25 °C

Description

Features and Benefits

Anionic polyelectrolyte. Dry form does not melt but decomposes at ca. 460C.
Despite water solubility, lithium form improves organic compatibility.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Articles

Polyelectrolyte Multilayer Films and Membrane Functionalization

Among the many methods for preparing thin films, layer-by-layer (LbL) deposition of complementary polymers has emerged as an especially versatile technique for controlling film thickness and function...
Merlin L. Bruening, Maneesha Adusumilli
Material Matters Volume 6 Number 3
Keywords: Adsorption, Catalysis, Deposition, Diffusion, Digestions, Electrophoresis, Gel electrophoresis, Immobilization, Mass spectrometry, Nanotechnology, Phase transitions, Reductions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?