• USA Home
  • 561916 - L-Tartaric acid, polymer-bound
EMAIL THIS PAGE TO A FRIEND
561916 Aldrich
L-Tartaric acid, polymer-bound
100-200 mesh, extent of labeling: 0.9-1.1 mmol/g loading, 1 % cross-linked with divinylbenzene
Purchase
Properties
Related Categories Chemical Synthesis, Other Supported Reagents, Solid Supported Synthesis, Specialty Synthesis, Supported Reagents More...
crosslinking   1 % cross-linked with divinylbenzene
extent of labeling   0.9-1.1 mmol/g loading
particle size   100-200 mesh
Price and Availability
Cross-Coupling Guide
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?