• USA Home
  • 562513 - Cesium hydroxide monohydrate

EMAIL THIS PAGE TO A FRIEND
562513 Aldrich

Cesium hydroxide monohydrate

technical grade

Purchase

Properties

Related Categories Acids & Bases, Bases, Cesium Salts, Chemical Synthesis, Essential Chemicals,
grade   technical grade
InChI Key   HUCVOHYBFXVBRW-UHFFFAOYSA-M
form   powder
mp   272 °C(lit.)
density   3.68 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2682 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?