• USA Home
  • 562963 - 3-Methyl-1-(4′-sulfoamidophenyl)-5-pyrazolone

EMAIL THIS PAGE TO A FRIEND
562963 Aldrich

3-Methyl-1-(4′-sulfoamidophenyl)-5-pyrazolone

95%

Purchase

Properties

Related Categories Building Blocks, C10, Chemical Synthesis, Heterocyclic Building Blocks, Pyrazoles More...
InChI Key   GLOOJKUWIGSFGC-UHFFFAOYSA-N
assay   95%
mp   246-250 °C(lit.)
Gene Information   human ... CA2(760)

Description

Packaging

25 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?