• USA Home
  • 562971 - 3-Methyl-1-(3′-sulfoamidophenyl)-5-pyrazolone

EMAIL THIS PAGE TO A FRIEND
562971 Aldrich

3-Methyl-1-(3′-sulfoamidophenyl)-5-pyrazolone

97%

Purchase

Properties

Related Categories Building Blocks, C10, Chemical Synthesis, Heterocyclic Building Blocks, Pyrazoles More...
InChI Key   ASVVGQURNHNITH-UHFFFAOYSA-N
assay   97%
mp   202-206 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?