• USA Home
  • 562998 - (3Z)-4-[(2-Hydroxyethyl)amino]-3-penten-2-one

EMAIL THIS PAGE TO A FRIEND
562998 Aldrich

(3Z)-4-[(2-Hydroxyethyl)amino]-3-penten-2-one

97%

Purchase

Properties

Related Categories Building Blocks, C7 to C8, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   CNRDGDCLFLAOFB-WAYWQWQTSA-N
assay   97%
mp   74-78 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?