• USA Home
  • 563005 - α-(4-Tolylimino)-p-cresol

EMAIL THIS PAGE TO A FRIEND
563005 Aldrich

α-(4-Tolylimino)-p-cresol

96%

Synonym: N-(4-Hydroxybenzylidene)-p-toluidine

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Imines/Amidines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   LXUWDLKAQOFUGU-UHFFFAOYSA-N
assay   96%
mp   218-222 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?