• USA Home
  • 563374 - Cyclopropylacetylene solution

EMAIL THIS PAGE TO A FRIEND
563374 Aldrich

Cyclopropylacetylene solution

70 wt. % in toluene

Synonym: Ethynylcyclopropane solution

Purchase

Properties

Related Categories Alkynes, Building Blocks, Chemical Synthesis, Organic Building Blocks, Terminal More...
InChI Key   NPTDXPDGUHAFKC-UHFFFAOYSA-N
concentration   70 wt. % in toluene
refractive index   n20/D 1.4480
bp   79-80 °C
bulk density   0.797 g/mL

Description

Packaging

100 mL in Sure/Seal™

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN1294 - class 3 - PG 2 - Toluene
WGK Germany 
2
Flash Point(F) 
-4 °F
Flash Point(C) 
-20 °C
Protocols & Articles

Related Content

Alkynes and Acetylenic Building Blocks

Aldrich Chemistry lists over 600 alkyne building blocks, including a wide array of propargyl alcohols, acetylenic boron reagents, and halogenated substrates for cross-coupling. Additionally, many of ...
Keywords: Building blocks, Coupling reactions, Cross couplings

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?