• USA Home
  • 563587 - 4-(Trifluoromethoxy)benzenesulfonamide

EMAIL THIS PAGE TO A FRIEND
563587 Aldrich

4-(Trifluoromethoxy)benzenesulfonamide

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonamides/Sulfinamides, Sulfur Compounds More...
InChI Key   RGOJCHYYBKMRLL-UHFFFAOYSA-N
assay   97%
mp   146-148 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?