• USA Home
  • 564850 - HMPB-benzhydrylamine, polymer-bound

EMAIL THIS PAGE TO A FRIEND
564850 Aldrich

HMPB-benzhydrylamine, polymer-bound

100-200 mesh, extent of labeling: 0.7-1.3 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym: 4-(4-Hydroxymethyl-3-methoxyphenoxy)butyryl-BHA-polystyrene, 4-Hydroxymethyl-3-methoxyphenoxybutyric acid, polymer-bound to BHA resin, HMPB-BHA resin

Purchase

Properties

Related Categories Chemical Synthesis, Other Resins, Solid Supported Synthesis, Specialty Synthesis
crosslinking   1 % cross-linked with divinylbenzene
extent of labeling   0.7-1.3 mmol/g loading
particle size   100-200 mesh

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?