• USA Home
  • 564869 - Bromoacetal, polymer-bound
EMAIL THIS PAGE TO A FRIEND
564869 Aldrich
Bromoacetal, polymer-bound
75-100 mesh, extent of labeling: ~1.5 mmol/g loading, 1 % cross-linked with divinylbenzene
Purchase
Properties
Related Categories Chemical Synthesis, Other Resins, Solid Supported Synthesis, Specialty Synthesis
crosslinking   1 % cross-linked with divinylbenzene
extent of labeling   ~1.5 mmol/g loading
particle size   75-100 mesh
Description
Packaging
5, 25 g in glass bottle
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?