• USA Home
  • 565083 - Poly[(methyl methacrylate)-co-(N-(1-naphthyl)-N-phenylacrylamide)]

EMAIL THIS PAGE TO A FRIEND
565083 Aldrich

Poly[(methyl methacrylate)-co-(N-(1-naphthyl)-N-phenylacrylamide)]

Purchase

Properties

Related Categories Fluorescent Polymers, Materials Science, Organic and Printed Electronics, Photoluminescent Materials, Photonic and Optical Materials More...
composition   N-(1-naphthyl)-N-phenylacrylamide, ~20 mol %
transition temp   Tg (DSC) 67 °C (onset)
fluorescence   λex 251 nm; λem 425 nm in chloroform

Description

Application

Fluorescent polymer.

Packaging

500 mg in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?