• USA Home
  • 565091 - N-(1-Naphthyl)-N-phenylmethacrylamide

EMAIL THIS PAGE TO A FRIEND
565091 Aldrich

N-(1-Naphthyl)-N-phenylmethacrylamide

98%

Purchase

Properties

Related Categories Fluorescent Monomers, Materials Science, Organic and Printed Electronics, Photoluminescent Materials, Photonic and Optical Materials More...
InChI Key   BLVLNPMGKHGHJN-UHFFFAOYSA-N
assay   98%
contains   500-1,000 ppm MEHQ as additive
mp   112-118 °C(lit.)
absorption   λmax 222 nm
storage temp.   2-8°C

Description

Packaging

100 mg in glass insert

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?