• USA Home
  • 566306 - (N,N-Dimethylamino)ethenyl-2,4-dinitrobenzene

EMAIL THIS PAGE TO A FRIEND
566306 Aldrich

(N,N-Dimethylamino)ethenyl-2,4-dinitrobenzene

≥97%

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   MHIRUPHZSNZDCI-AATRIKPKSA-N
assay   ≥97%
mp   163-167 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?