• USA Home
  • 566454 - 3-(Difluoromethoxy)benzonitrile

EMAIL THIS PAGE TO A FRIEND
566454 Aldrich

3-(Difluoromethoxy)benzonitrile

98%

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   JCBOPIBHZFBHQW-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.4870(lit.)
bp   224-225 °C(lit.)
density   1.260 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?