• USA Home
  • 566497 - 1-[3,5-Di-(trifluoromethyl)phenyl]biguanide hydrochloride

EMAIL THIS PAGE TO A FRIEND
566497 Aldrich

1-[3,5-Di-(trifluoromethyl)phenyl]biguanide hydrochloride

  • CAS Number 36068-40-3

  • Linear Formula (CF3)2C6H3NHC(=NH)NHC(=NH)NH2 · HCl

  • Molecular Weight 349.66

  •  MDL number MFCD03094450

  •  PubChem Substance ID 24880301

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Guanidines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   VEHPYWMGWYCGTE-UHFFFAOYSA-N
mp   246 °C (dec.)(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?