• USA Home
  • 566500 - 1-[4-(Trifluoromethoxy)phenyl]biguanide hydrochloride

EMAIL THIS PAGE TO A FRIEND
566500 Aldrich

1-[4-(Trifluoromethoxy)phenyl]biguanide hydrochloride

97%

  • CAS Number 42823-09-6

  • Linear Formula CF3OC6H4NHC(=NH)NHC(=NH)NH2 ·HCl

  • Molecular Weight 297.66

  •  MDL number MFCD01569536

  •  PubChem Substance ID 24880303

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Guanidines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   VAWGZGSCMHDUCU-UHFFFAOYSA-N
assay   97%
mp   207-212 °C(lit.)

Description

Packaging

1, 5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?