• USA Home
  • 566519 - 1-(3-Fluorophenyl)biguanide hydrochloride

EMAIL THIS PAGE TO A FRIEND
566519 Aldrich

1-(3-Fluorophenyl)biguanide hydrochloride

97%

  • CAS Number 2267-49-4

  • Linear Formula FC6H4NHC=NHNHC=NHNH2 · HCl

  • Molecular Weight 231.66

  •  MDL number MFCD00203529

  •  PubChem Substance ID 24880304

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Guanidines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   IWYOYHXLNBELKJ-UHFFFAOYSA-N
assay   97%
mp   231-235 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?