• USA Home
  • 56657 - (+)-Methyl (S)-3-hydroxyvalerate

EMAIL THIS PAGE TO A FRIEND
56657 Aldrich

(+)-Methyl (S)-3-hydroxyvalerate

≥98.0% (sum of enantiomers, GC)

Synonym: (+)-Methyl (S)-3-hydroxypentanoate

  • CAS Number 42558-50-9

  • Empirical Formula (Hill Notation) C6H12O3

  • Molecular Weight 132.16

  •  Beilstein Registry Number 4655381

  •  MDL number MFCD00216722

  •  PubChem Substance ID 24880308

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Esters, Organic Building Blocks More...
InChI Key   XHFXKKFVUDJSPJ-YFKPBYRVSA-N
assay   ≥98.0% (sum of enantiomers, GC)
optical activity   [α]20/D +37±2°, c = 1% in chloroform
refractive index   n20/D 1.427
density   1.029 g/mL at 20 °C(lit.)

Description

Caution

may discolor to brown on storage

Packaging

1, 5 mL in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?