• USA Home
  • 566799 - Benzophenone tosylhydrazone

EMAIL THIS PAGE TO A FRIEND
566799 Aldrich

Benzophenone tosylhydrazone

97%

Synonym: Toluene-4-sulfonic acid benzhydrylidenehydrazone

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Hydrazones, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   AVWHDUALELPLAI-UHFFFAOYSA-N
assay   97%
mp   178-182 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?