• USA Home
  • 566837 - Valerophenone tosylhydrazone

EMAIL THIS PAGE TO A FRIEND
566837 Aldrich

Valerophenone tosylhydrazone

mixture of isomers

  • CAS Number 69015-74-3

  • Linear Formula CH3C6H4SO2NHN=C(C6H5)(CH2)3CH3

  • Molecular Weight 330.44

  •  MDL number MFCD00159338

  •  PubChem Substance ID 24880328

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Hydrazones, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   VTEOESQGMPJHGP-VHEBQXMUSA-N
mp   132-136 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?