EMAIL THIS PAGE TO A FRIEND
566888 Aldrich

1-Allylpiperazine

96%

Purchase

Properties

Related Categories Building Blocks, C4 to C8, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
InChI Key   ZWAQJGHGPPDZSF-UHFFFAOYSA-N
assay   96%
refractive index   n20/D 1.4830(lit.)
bp   181-182 °C(lit.)
density   0.902 g/mL(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2734PSN1 3(8) / PGII
WGK Germany 
3
Flash Point(F) 
123.8 °F
Flash Point(C) 
51 °C
Protocols & Articles

Articles

Piperazines

Sigma-Aldrich is pleased to offer the third installment of our Privileged Structures ChemFiles. This piece will focus on the piperazine scaffold. We will discuss application data, Sigma-Aldrich build...
Aldrich ChemFiles 2005, 5.5, 1.
Keywords: Anti-depressants, Antifungals, Antivirals, Building blocks, Chemfiles, Company, Medicinal chemistry, Metabolism, Organic synthesis, Pharmaceutical

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?