• USA Home
  • 567329 - 2-Ethoxy-3-formyl-5-methylphenylboronic acid

EMAIL THIS PAGE TO A FRIEND
567329 Aldrich

2-Ethoxy-3-formyl-5-methylphenylboronic acid

Purchase

Description

Other Notes

Contains varying amounts of anhydride

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1759 8 / PGIII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?