• USA Home
  • 567825 - 1-Isocyanato-1,2,3,4-tetrahydronaphthalene

EMAIL THIS PAGE TO A FRIEND
567825 Aldrich

1-Isocyanato-1,2,3,4-tetrahydronaphthalene

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Isocyanates, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   OABBXWCERFBLEZ-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.5470(lit.)
bp   268 °C(lit.)
density   1.104 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Packaging

1, 5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 3334
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?