• USA Home
  • 567892 - 2,3-Dimethoxyphenethyl isocyanate

EMAIL THIS PAGE TO A FRIEND
567892 Aldrich

2,3-Dimethoxyphenethyl isocyanate

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Isocyanates, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   TWNQNDPGGHUEGM-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.5250(lit.)
bp   275-276 °C(lit.)
density   1.131 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Caution

Refrigerate

Packaging

1, 5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 2206PSN1B 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?