• USA Home
  • 567922 - 5-Bromo-2-methoxyphenethyl isocyanate

EMAIL THIS PAGE TO A FRIEND
567922 Aldrich

5-Bromo-2-methoxyphenethyl isocyanate

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Isocyanates, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   IEKFZVSKEVFNQZ-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.5620(lit.)
bp   280-281 °C(lit.)
density   1.454 g/mL at 25 °C(lit.)

Description

Caution

Refrigerate

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?