• USA Home
  • 569860 - 3-(1,3,4,6,7,8,-Hexahydro-2H-pyrimido-[1,2-a]pyrimidino)propyl, functionalized silica gel

EMAIL THIS PAGE TO A FRIEND
569860 Aldrich

3-(1,3,4,6,7,8,-Hexahydro-2H-pyrimido-[1,2-a]pyrimidino)propyl, functionalized silica gel

200-400 mesh, extent of labeling: 1.0 mmol/g loading

Synonym: 1,5,7-Triazabicyclo[4.4.0]dec-5-ene, silica gel supported, TBD on silica gel

Purchase

Properties

Related Categories Chemical Synthesis, Reagents, Silica Gel, Solid Supported Synthesis, Specialty Synthesis More...
extent of labeling   1.0 mmol/g loading
particle size   200-400 mesh
pore size   60 Å pore size
surface area   500 m2/g
storage temp.   2-8°C

Description

Packaging

5, 25 g in glass bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?