• USA Home
  • 569909 - 3-(Maleimido)propyl, functionalized silica gel

EMAIL THIS PAGE TO A FRIEND
569909 Aldrich

3-(Maleimido)propyl, functionalized silica gel

200-400 mesh, extent of labeling: 0.7 mmol/g loading

Synonym: Maleimide, silica gel supported

Purchase

Properties

Related Categories Chemical Synthesis, Scavengers, Silica Gel, Solid Supported Synthesis, Specialty Synthesis More...
extent of labeling   0.7 mmol/g loading
particle size   200-400 mesh
pore size   60 Å pore size
surface area   500 m2/g
storage temp.   2-8°C

Description

Application

Scavenges thiols

Packaging

5 g in glass bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?