• USA Home
  • 570451 - Disperse Red 13 methacrylate

EMAIL THIS PAGE TO A FRIEND
570451 Aldrich

Disperse Red 13 methacrylate

98%

Purchase

Properties

Related Categories Materials Science, NLO Monomers, Non-Linear Optical (NLO) Materials, Organic and Printed Electronics, Photonic and Optical Materials More...
InChI Key   UJXGDBTUUWCZLZ-GHVJWSGMSA-N
assay   98%
mp   66-71 °C(lit.)
absorption   λmax 204 nm
storage temp.   2-8°C

Description

General description

NLO material

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?