• USA Home
  • 570613 - (4R)-4-(Chloromethyl)-3-[(1R)-1-phenylethyl]-2-oxazolidinone

EMAIL THIS PAGE TO A FRIEND
570613 Aldrich

(4R)-4-(Chloromethyl)-3-[(1R)-1-phenylethyl]-2-oxazolidinone

98%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Auxiliaries, Oxazolidinone Derivatives
InChI Key   RDJVMESMFKYWJT-KOLCDFICSA-N
assay   98%
optical purity   ee: 98% (GLC)
mp   60-65 °C(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?