• USA Home
  • 571210 - [3-(1-Piperidinylmethyl)phenyl]magnesium bromide solution

EMAIL THIS PAGE TO A FRIEND
571210 Aldrich

[3-(1-Piperidinylmethyl)phenyl]magnesium bromide solution

0.25 M in THF

Purchase

Properties

Related Categories Aryl, Chemical Synthesis, Grignard Reagents, Organometallic Reagents
InChI Key   QLVYNMWCYBTNEP-UHFFFAOYSA-M
concentration   0.25 M in THF
bp   65 °C
density   0.919 g/mL at 25 °C
storage temp.   2-8°C

Description

Packaging

50 mL in Sure/Seal™

Legal Information

Product of Rieke® Metals, Inc.

Rieke is a registered trademark of Rieke Metals, Inc.

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
May form explosive peroxides., Reacts violently with water.
RIDADR 
UN 2924 8(3) / PGII
WGK Germany 
3
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?