• USA Home
  • 571369 - (4-Bromophenylthio)dimethyl-tert-butylsilane

EMAIL THIS PAGE TO A FRIEND
571369 Aldrich

(4-Bromophenylthio)dimethyl-tert-butylsilane

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Other Sulfur Compounds, Sulfur Compounds More...
InChI Key   ZVSDFKXSTADQGI-UHFFFAOYSA-N
assay   97%
bp   120-130 °C/0.2-0.3 mmHg(lit.)
mp   42-44 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?