• USA Home
  • 575909 - trans-Dichlorobis(ethylenediamine)cobalt(III) chloride

EMAIL THIS PAGE TO A FRIEND
575909 Aldrich

trans-Dichlorobis(ethylenediamine)cobalt(III) chloride

99.999% trace metals basis

  • Empirical Formula (Hill Notation) C4H16Cl3CoN4

  • Molecular Weight 285.49

  •  MDL number MFCD00054234

  •  PubChem Substance ID 329758324

Purchase

Properties

Related Categories Cobalt Salts, Electronic Chemicals, Essential Chemicals, Materials Science, Metal and Ceramic Science,
InChI Key   GVMSQWCTZLHSQH-UHFFFAOYSA-K
assay   99.999% trace metals basis
form   powder and chunks
mp   237-239 °C (dec.)(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?