• USA Home
  • 58923 - (1R)-(+)-trans-Isolimonene

EMAIL THIS PAGE TO A FRIEND
58923 Aldrich

(1R)-(+)-trans-Isolimonene

≥95.0% (sum of enantiomers, GC)

Synonym: (+)-p-Mentha-2,8-diene, (3R,6R)-3-Isopropenyl-6-methylcyclohexene

  • CAS Number 5113-87-1

  • Empirical Formula (Hill Notation) C10H16

  • Molecular Weight 136.23

  •  Beilstein Registry Number 2073245

  •  EC Number 225-843-3

  •  MDL number MFCD00042629

  •  PubChem Substance ID 24881064

Purchase

Properties

Related Categories Alkenes, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   TWCNAXRPQBLSNO-UWVGGRQHSA-N
assay   ≥95.0% (sum of enantiomers, GC)
optical activity   [α]20/D +212±5°, c = 10% in ethanol
refractive index   n20/D 1.47(lit.)
bp   165-166 °C(lit.)
density   0.83 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
RIDADR 
UN 2052 3 / PGIII
WGK Germany 
3
Flash Point(F) 
102.2 °F
Flash Point(C) 
39 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?