• USA Home
  • 58926 - (−)-Isolongifolen-9-one

EMAIL THIS PAGE TO A FRIEND
58926 Aldrich

(−)-Isolongifolen-9-one

≥99.5% (sum of enantiomers, GC)

Synonym: (1R)-2,2,7,7-Tetramethyltricyclo[6.2.1.01,6]undec-5-en-4-one

  • CAS Number 26839-52-1

  • Empirical Formula (Hill Notation) C15H22O

  • Molecular Weight 218.33

  •  Beilstein Registry Number 2213328

  •  MDL number MFCD00270137

  •  PubChem Substance ID 24881068

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Ketones, Organic Building Blocks More...
InChI Key   LGSKOQUJWNADCQ-BONVTDFDSA-N
assay   ≥99.5% (sum of enantiomers, GC)
optical activity   [α]20/D −205±10°, c = 1% in ethanol
bp   143-144 °C/4.5 mmHg(lit.)
mp   34-37 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?