EMAIL THIS PAGE TO A FRIEND
59059 Aldrich

Luviquat HM 552

~20% active ingredients in H2O

Synonym: Poly[(3-methyl-1-vinylimidazolium chloride)-co-(1-vinylpyrrolidone)], Polyquaternium D10

Purchase

Properties

Related Categories Biochemicals and Reagents, Cationic Detergents, Detergents, Detergents A to M
description   cationic
concentration   ~20% active ingredients in H2O
refractive index   n20/D 1.372
pH   4.5-7.5 (10% in H2O, active ingredients)
density   1.05 g/mL at 20 °C

Description

Legal Information

Luviquat is a trademark of BASF SE

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?