• USA Home
  • 59442 - (S)-4-Benzyloxymethyl-2,2-dimethyl-1,3-dioxolane

EMAIL THIS PAGE TO A FRIEND
59442 Aldrich

(S)-4-Benzyloxymethyl-2,2-dimethyl-1,3-dioxolane

≥95.0% (sum of enantiomers, GC)

Synonym: 3-Benzyl-1,2-isopropylidene-sn-glycerol, D-α,β-Isopropylideneglycerol-γ-benzyl ether

  • CAS Number 16495-03-7

  • Empirical Formula (Hill Notation) C13H18O3

  • Molecular Weight 222.28

  •  Beilstein Registry Number 82963

  •  MDL number MFCD00043000

  •  PubChem Substance ID 57651684

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Protected Alcohols More...
InChI Key   DBFDSKSLTCMIPB-LBPRGKRZSA-N
assay   ≥95.0% (sum of enantiomers, GC)
optical activity   [α]20/D +21±1°, c = 4% in chloroform
refractive index   n20/D 1.496
bp   79-80 °C/0.05 mmHg(lit.)
density   1.050 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Chiral building block; synthesis of chiral glycerides

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?