• USA Home
  • 59814 - tert-Butyl 3-isothiocyanatobenzoate

EMAIL THIS PAGE TO A FRIEND
59814 Aldrich

tert-Butyl 3-isothiocyanatobenzoate

≥97.0% (HPLC)

Synonym: 3-(tert-Butoxycarbonyl)phenyl isothiocyanate

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiocyanates/Isothiocyanates More...
InChI Key   WUMCWUULWWVQGT-UHFFFAOYSA-N
assay   ≥97.0% (HPLC)
mp   37-40 °C

Description

Packaging

1 g in poly bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?