EMAIL THIS PAGE TO A FRIEND
59870 Aldrich

Isobutyl isovalerate

≥98.0% (GC)

Synonym: 2-Methylpropyl 3-methylbutyrate, Isobutyl 3-methyl butyrate

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Carbonyl Compounds, Cell Biology, Chamaemelum nobile (Chamomile tea),
InChI Key   KEBDNKNVCHQIJU-UHFFFAOYSA-N
assay   ≥98.0% (GC)
refractive index   n20/D 1.406

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
NY1506000
Flash Point(F) 
127.4 °F
Flash Point(C) 
53 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?