EMAIL THIS PAGE TO A FRIEND
62861 Aldrich

Sodium borohydride

≥95%

Synonym: Sodium tetrahydridoborate

Purchase

Properties

Related Categories Borohydrides, Chemical Synthesis, Reduction, Synthetic Reagents
InChI Key   YOQDYZUWIQVZSF-UHFFFAOYSA-N
assay   ≥95%
mp   >300 °C (dec.)(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 1426 4.3 / PGI
WGK Germany 
2
RTECS 
ED3325000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

452882 Sodium borohydride, powder, ≥98.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?