Synonym: Heavy water, Water-d2
CAS Number 7789-20-0
Empirical Formula (Hill Notation) D2O
Molecular Weight 20.03
EC Number 232-148-9
MDL number MFCD00044636
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, D, DA - DH, |
| isotopic purity | 99.994 atom % D |
| contains | 0.01 % (w/v) DSS-d6 |
| 1 mM terephthalic acid disodium salt | |
| refractive index |
n |
| bp | 101.4 °C(lit.) |
| 101.4 °C | |
| mp | 3.8 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?